2-Bromo-4'-(4-morpholino)acetophenone 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
2-Bromo-1-[4-(morpholin-4-yl)phenyl]ethan-1-one
OR8584-1G
OR8584-250MG
OR8584-5G
APOSOR8584-1GEA
4670
CZK
APOSOR8584-1G
APOSOR8584-250MG
APOSOR8584-5G
2-Bromo-4'-(4-morpholino)acetophenone 97%
2-Bromo-4'-(4-morpholino)acetophenone
Vzorec:
C₁₂H₁₄BrNO₂ Bod tání: 112…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD03783555 CAS číslo: 210832-85-2 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...