6-Methylisoquinoline 98%
Dodavatel: Apollo Scientific
|
Varování
|
OR8565-1G
OR8565-250MG
OR8565-5G
APOSOR8565-1GEA
2320
CZK
APOSOR8565-1G
APOSOR8565-250MG
APOSOR8565-5G
6-Methylisoquinoline 98%
6-Methylisoquinoline
Vzorec:
C₁₀H₉N Storage Temperature: Ambient |
MDL Number:
MFCD09258767 CAS číslo: 42398-73-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...