Tisk…

8-Isoquinolinol 95%

Dodavatel: Apollo Scientific
Varování

Synonyms: 8-Hydroxyisoquinoline, Isoquinolin-8-ol

OR8561-1G OR8561-250MG OR8561-5G
APOSOR8561-1GEA 1910 CZK
APOSOR8561-1G APOSOR8561-250MG APOSOR8561-5G
8-Isoquinolinol 95%
8-Isoquinolinol
Vzorec: C₉H₇NO
MW: 145,1600 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00661583
CAS číslo: 3482-14-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR