Tisk…

3-Amino-5-chlorobenzonitrile 98%

Dodavatel: Apollo Scientific
Varování

OR8559-1G OR8559-5G OR8559-10G
APOSOR8559-1GEA 992 CZK
APOSOR8559-1G APOSOR8559-5G APOSOR8559-10G
3-Amino-5-chlorobenzonitrile 98%
3-Amino-5-chlorobenzonitrile
Vzorec: C₇H₅ClN₂
Storage Temperature: Ambient
MDL Number: MFCD06797224
CAS číslo: 53312-78-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR