Tisk…

(5-Bromopyrimidin-2-yl)piperidine 98%

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Bromo-2-(piperidin-1-yl)pyrimidine

OR8548-25G OR8548-5G OR8548-1G
APOSOR8548-25GEA 5200 CZK
APOSOR8548-25G APOSOR8548-5G APOSOR8548-1G
(5-Bromopyrimidin-2-yl)piperidine 98%
(5-Bromopyrimidin-2-yl)piperidine
Vzorec: C₉H₁₂BrN₃
MW: 242,1157 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00483257
CAS číslo: 57356-64-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR