[1-(4-Methylbenzyl)-1H-1,2,3-triazol-4-yl]methanol
Dodavatel: Apollo Scientific
OR8513-1G
OR8513-500MG
APOSOR8513-1GEA
9560
CZK
APOSOR8513-1G
APOSOR8513-500MG
[1-(4-Methylbenzyl)-1H-1,2,3-triazol-4-yl]methanol
[1-(4-Methylbenzyl)-1H-1,2,3-triazol-4-yl]methanol
Vzorec:
C₁₁H₁₃N₃O Bod tání: 85…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD09607911 CAS číslo: 952183-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...