α-Bromo-4-chlorotoulene 98%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
Synonyms:
alpha-Bromo-4-chlorotoluene, 1-(Bromomethyl)-4-chlorobenzene, 4-Chlorobenzyl bromide
OR8476-25G
OR8476-100G
OR8476-5G
APOSOR8476-25GEA
787
CZK
APOSOR8476-25G
APOSOR8476-100G
APOSOR8476-5G
α-Bromo-4-chlorotoulene 98%
α-Bromo-4-chlorotoulene
Vzorec:
C₇H₆BrCl MW: 205,48 g/mol Bod varu: 107…108 °C/8mmHg Bod tání: 46…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040714 CAS číslo: 622-95-7 EINECS: 210-760-7 UN: 3261 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...