1-(4-Nitrophenyl)-4-piperidinecarboxylic acid
Dodavatel: Apollo Scientific
OR8465-10G
OR8465-1G
OR8465-5G
APOSOR8465-10GEA
30220
CZK
APOSOR8465-10G
APOSOR8465-1G
APOSOR8465-5G
1-(4-Nitrophenyl)-4-piperidinecarboxylic acid
1-(4-Nitrophenyl)-4-piperidinecarboxylic acid
Vzorec:
C₁₂H₁₄N₂O₄ Bod tání: 200…201 °C Storage Temperature: Ambient |
CAS číslo:
223786-53-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...