5-Bromo-2-methox-4-methylypyridine 95%
Dodavatel: Apollo Scientific
OR8427-10G
OR8427-5G
APOSOR8427-10GEA
4340
CZK
APOSOR8427-10G
APOSOR8427-5G
5-Bromo-2-methox-4-methylypyridine 95%
5-Bromo-2-methox-4-methylypyridine
Vzorec:
C₇H₈BrNO Storage Temperature: Ambient |
MDL Number:
MFCD04039980 CAS číslo: 164513-39-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...