1-Propylboronic acid 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR8415-1G
OR8415-25G
OR8415-10G
APOSOR8415-1GEA
622
CZK
APOSOR8415-1G
APOSOR8415-25G
APOSOR8415-10G
1-Propylboronic acid 97%
1-Propylboronic acid
Vzorec:
C₃H₉BO₂ Bod tání: 105…108 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01074564 CAS číslo: 17745-45-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...