Tisk…

1H-Pyrazole-5-boronic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 5-Borono-1H-pyrazole, 2H-Pyrazole-3-boronic acid, (1H-Pyrazol-5-yl)boronic acid

OR8413-1G OR8413-10G OR8413-5G
APOSOR8413-1GEA 1490 CZK
APOSOR8413-1G APOSOR8413-10G APOSOR8413-5G
1H-Pyrazole-5-boronic acid
1H-Pyrazole-5-boronic acid
Vzorec: C₃H₅BN₂O₂
MW: 111,89 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD07368247
CAS číslo: 376584-63-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR