(2-Chloro-1,3-thiazol-5-yl)methanol
Dodavatel: Apollo Scientific
|
Varování
|
OR8406-1G
OR8406-5G
APOSOR8406-1GEA
1870
CZK
APOSOR8406-1G
APOSOR8406-5G
(2-Chloro-1,3-thiazol-5-yl)methanol
(2-Chloro-1,3-thiazol-5-yl)methanol
Vzorec:
C₄H₄NOClS Bod varu: 96 °C/0,2mmHg Storage Temperature: Refrigerator |
MDL Number:
MFCD03001277 CAS číslo: 145015-15-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...