Tisk…

3-Bromo-N,N-dimethyl-5-nitrobenzamide 98%

Dodavatel: Apollo Scientific

OR8307-1G OR8307-5G
APOSOR8307-1GEA 3940 CZK
APOSOR8307-1G APOSOR8307-5G
3-Bromo-N,N-dimethyl-5-nitrobenzamide 98%
3-Bromo-N,N-dimethyl-5-nitrobenzamide
Vzorec: C₉H₉BrN₂O₃
Storage Temperature: Ambient
MDL Number: MFCD09258761
CAS číslo: 929000-26-0

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR