3-Bromo-N,N-diethyl-5-nitrobenzamide 98%
Dodavatel: Apollo Scientific
OR8296-1G
OR8296-5G
APOSOR8296-1GEA
3740
CZK
APOSOR8296-1G
APOSOR8296-5G
3-Bromo-N,N-diethyl-5-nitrobenzamide 98%
3-Bromo-N,N-diethyl-5-nitrobenzamide
Vzorec:
C₁₁H₁₃BrN₂O₃ Storage Temperature: Ambient |
MDL Number:
MFCD09258763 CAS číslo: 929000-34-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...