Tisk…

7-Methoxy-4-quinolinol

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-Hydroxy-7-methoxyquinoline

OR8294-100G OR8294-25G OR8294-5G
APOSOR8294-100GEA 10020 CZK
APOSOR8294-100G APOSOR8294-25G APOSOR8294-5G
7-Methoxy-4-quinolinol
7-Methoxy-4-quinolinol
Vzorec: C₁₀H₉NO₂
MW: 175,19 g/mol
Bod tání: 213…217 °C
Storage Temperature: Ambient
MDL Number: MFCD00169015
CAS číslo: 82121-05-9
UN: 3335
ADR: 9,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR