4-Nitrophthalonitrile
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Nitrobenzene-1,2-dicarbonitrile, 4-Nitro-1,2-benzenedicarbonitrile, 4-Nitro-1,2-dicyanobenzene
OR8247-100G
OR8247-25G
OR8247-5G
OR8247-500G
OR8247-1KG
APOSOR8247-100GEA
953
CZK
APOSOR8247-100G
APOSOR8247-25G
APOSOR8247-5G
APOSOR8247-500G
APOSOR8247-1KG
4-Nitrophthalonitrile
4-Nitrophthalonitrile
Vzorec:
C₈H₃N₃O₂ MW: 173,13 g/mol Bod tání: 136…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040301 CAS číslo: 31643-49-9 EINECS: 250-748-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...