Tisk…

1,2,3-Thiadiazole-4-carbaldehyde 97%

Dodavatel: Apollo Scientific

OR8149-1G OR8149-250MG
APOSOR8149-1GEA 8670 CZK
APOSOR8149-1G APOSOR8149-250MG
1,2,3-Thiadiazole-4-carbaldehyde 97%
1,2,3-Thiadiazole-4-carbaldehyde
Vzorec: C₃H₂N₂OS
MDL Number: MFCD02180762
CAS číslo: 27643-15-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR