Tisk…

Carboxy-PTIO potassium salt

Dodavatel: Apollo Scientific

OR8125T-10MG OR8125T-50MG
APOSOR8125T-10MGEA 0 CZK
APOSOR8125T-10MG APOSOR8125T-50MG
Carboxy-PTIO potassium salt
Carboxy-PTIO potassium salt

A water soluble, stable free radical that reacts stoichiometrically with NO.

Vzorec: C₁₄H₁₆KN₂O₄
MW: 315,4 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00216153
CAS číslo: 148819-94-7

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR