Tisk…

5-[(Dimethylamino)sulphonyl]-2-methyl-3-furoic acid 97%

Dodavatel: Apollo Scientific
Nebezpečí

OR8125-1G OR8125-250MG
APOSOR8125-1GEA 3110 CZK
APOSOR8125-1G APOSOR8125-250MG
5-[(Dimethylamino)sulphonyl]-2-methyl-3-furoic acid 97%
5-[(Dimethylamino)sulphonyl]-2-methyl-3-furoic acid
Vzorec: C₈H₁₁NO₅S
MW: 233,24 g/mol
Bod tání: 169 °C
Storage Temperature: Ambient
MDL Number: MFCD02180761
CAS číslo: 306936-39-0
UN: 3261
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR