2-Isobutylthiazole 98%
Dodavatel: Apollo Scientific
OR8109-100G
OR8109-25G
OR8109-5G
APOSOR8109-100GEA
12870
CZK
APOSOR8109-100G
APOSOR8109-25G
APOSOR8109-5G
2-Isobutylthiazole 98%
2-Isobutylthiazole
Vzorec:
C₇H₁₁NS MW: 141,24 g/mol Bod varu: 175 °C/760mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00005334 CAS číslo: 18640-74-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...