2-(4-Isobutylphenyl)propanenitrile 97%
Dodavatel: Apollo Scientific
|
Varování
|
OR8108-1G
OR8108-5G
APOSOR8108-1GEA
5200
CZK
APOSOR8108-1G
APOSOR8108-5G
2-(4-Isobutylphenyl)propanenitrile 97%
2-(4-Isobutylphenyl)propanenitrile
Vzorec:
C₁₃H₁₇N Bod varu: 124 °C/0,8mmHg Storage Temperature: Ambient |
MDL Number:
MFCD02180706 CAS číslo: 58609-73-7 EINECS: 261-365-1 UN: 3276 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...