Tisk…

6-(tert-Butyl)-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone

Dodavatel: Apollo Scientific
Varování

Synonyms: 4-tert-Butyl-6-hydroxy-2-mercaptopyrimidine

OR8097-1G OR8097-250MG
APOSOR8097-1GEA 6650 CZK
APOSOR8097-1G APOSOR8097-250MG
6-(tert-Butyl)-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone
6-(tert-Butyl)-2-thioxo-2,3-dihydro-4(1H)-pyrimidinone
Vzorec: C₈H₁₂N₂OS
MW: 184,26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00466456
CAS číslo: 66698-66-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR