Tisk…

Phenylguanidine nitrate 99%

Dodavatel: Apollo Scientific
Varování

OR8070-10G
APOSOR8070-10GEA 7610 CZK
APOSOR8070-10G
Phenylguanidine nitrate 99%
Phenylguanidine nitrate
Vzorec: C₇H₁₀N₄O₃
MW: 198,18 g/mol
Bod tání: 122…124 °C
Storage Temperature: Ambient
MDL Number: MFCD00213415
CAS číslo: 18860-78-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR