5-Chloro-1-methyl-1H-pyrazole-4-carboxylic acid 94%
Dodavatel: Apollo Scientific
|
Varování
|
OR8059-10G
OR8059-1G
OR8059-5G
APOSOR8059-10GEA
3870
CZK
APOSOR8059-10G
APOSOR8059-1G
APOSOR8059-5G
5-Chloro-1-methyl-1H-pyrazole-4-carboxylic acid 94%
5-Chloro-1-methyl-1H-pyrazole-4-carboxylic acid
Vzorec:
C₅H₅ClN₂O₂ Bod tání: 197…198 °C Storage Temperature: Ambient |
MDL Number:
MFCD02089694 CAS číslo: 54367-66-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...