Tisk…

1-Boc-4-(aminomethyl)piperidine 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-(Aminomethyl)-1-tert-butoxycarbonylpiperidine, tert-Butyl 4-(aminomethyl)piperidine-1-carboxylate, tert-Butyl 4-(Aminomethyl)-1-piperidinecarboxylate, tert-Butyl 4-(aminomethyl)tetrahydropyridine-1(2H)- carboxylate, 4-(Aminomethyl)-1-Boc-piperidine, 4-(Aminomethyl)-1-piperidinecarboxylic acid tert-Butyl ester, 1-Boc-4-(Aminomethyl)piperidine

OR8043-1G OR8043-25G OR8043-5G
APOSOR8043-1GEA 622 CZK
APOSOR8043-1G APOSOR8043-25G APOSOR8043-5G
1-Boc-4-(aminomethyl)piperidine 97%
1-Boc-4-(aminomethyl)piperidine
Vzorec: C₁₁H₂₂N₂O₂
MW: 214,31 g/mol
Bod varu: 237…238 °C/760mmHg
Hustota: 1,013 g/cm³ (25 °C)
Bod vzplanutí: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD01076207
CAS číslo: 144222-22-0
UN: 3082
ADR: 9+EHS,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR