2-Bromo-3-methylthiophene 97%
Dodavatel: Apollo Scientific
|
Nebezpečí
|
OR8041-25G
OR8041-1G
APOSOR8041-25GEA
622
CZK
APOSOR8041-25G
APOSOR8041-1G
2-Bromo-3-methylthiophene 97%
2-Bromo-3-methylthiophene
Vzorec:
C₅H₅BrS MW: 177.06 Bod varu: 173…176 °C Hustota: 1.58 g/cm³ (25 °C) Bod vzplanutí: 68 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00059741 CAS číslo: 14282-76-9 EINECS: 238-175-2 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...