Tisk…

2-(1,2-Benzisoxazol-3-yl)acetic acid 97%

Dodavatel: Apollo Scientific
Varování

Synonyms: 1,2-Benzisoxazole-3-acetic Acid

OR8018-100G OR8018-1G
APOSOR8018-100GEA 5130 CZK
APOSOR8018-100G APOSOR8018-1G
2-(1,2-Benzisoxazol-3-yl)acetic acid 97%
2-(1,2-Benzisoxazol-3-yl)acetic acid
Vzorec: C₉H₇NO₃
MW: 177,1568 g/mol
Bod tání: 123…129 °C
Storage Temperature: Ambient
MDL Number: MFCD02180388
CAS číslo: 4865-84-3

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR