(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Chloropurine riboside
OR7990T-5G
OR7990T-25G
APOSOR7990T-5GEA
787
CZK
APOSOR7990T-5G
APOSOR7990T-25G
(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
(2R,3R,4S,5R)-2-(6-Chloro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol
A substrate for the enzyme adenosine deaminase.
Vzorec:
C₁₀H₁₁ClN₄O₄ MW: 286,67 g/mol Bod tání: 158…162 °C Storage Temperature: Freezer |
MDL Number:
MFCD00005738 CAS číslo: 5399-87-1 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...