Pyrimidine-4-carbaldehyde 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
4-Formylpyrimidine, 4-Formyl-1,3-diazine, 4-Pyrimidinecarboxaldehyde
OR7987-1G
OR7987-250MG
OR7987-5G
APOSOR7987-1GEA
7670
CZK
APOSOR7987-1G
APOSOR7987-250MG
APOSOR7987-5G
Pyrimidine-4-carbaldehyde 95%
Pyrimidine-4-carbaldehyde
Vzorec:
C₅H₄N₂O MW: 108.1 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03789624 CAS číslo: 2435-50-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...