1,2-Dibromo-4-(methylsulphonyl)benzene 95+%
Dodavatel: Apollo Scientific
OR7977-1G
OR7977-5G
OR7977-25G
APOSOR7977-1GEA
3640
CZK
APOSOR7977-1G
APOSOR7977-5G
APOSOR7977-25G
1,2-Dibromo-4-(methylsulphonyl)benzene 95+%
1,2-Dibromo-4-(methylsulphonyl)benzene
Vzorec:
C₇H₆Br₂O₂S Bod tání: 141…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD04037933 CAS číslo: 849035-70-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...