Tisk…

3,5-Bis(methylsulphonyl)benzonitrile 95+%

Dodavatel: Apollo Scientific

OR7953-1G OR7953-5G OR7953-25G
APOSOR7953-1GEA 5920 CZK
APOSOR7953-1G APOSOR7953-5G APOSOR7953-25G
3,5-Bis(methylsulphonyl)benzonitrile 95+%
3,5-Bis(methylsulphonyl)benzonitrile
Vzorec: C₉H₉NO₄S₂
Storage Temperature: Ambient
MDL Number: MFCD04037977
CAS číslo: 849924-84-1

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR