Tisk…

3,5-Bis(methylsulphonyl)benzenesulphonylchloride 95+%

Dodavatel: Apollo Scientific
Nebezpečí

OR7952-1G OR7952-5G
APOSOR7952-1GEA 5920 CZK
APOSOR7952-1G APOSOR7952-5G
3,5-Bis(methylsulphonyl)benzenesulphonylchloride 95+%
3,5-Bis(methylsulphonyl)benzenesulphonylchloride
Vzorec: C₈H₉ClO₆S₃
Storage Temperature: Ambient
MDL Number: MFCD04037976
CAS číslo: 849035-99-0
UN: 1759
ADR: 8,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR