2-Oxocyclohexanecarbonitrile
Dodavatel: Apollo Scientific
|
Varování
|
OR7944-1G
APOSOR7944-1GEA
1660
CZK
APOSOR7944-1G
2-Oxocyclohexanecarbonitrile
2-Oxocyclohexanecarbonitrile
Vzorec:
C₇H₉NO MW: 123,1525 g/mol Bod varu: 150 °C/20mmHg Hustota: 1,0741 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00154839 CAS číslo: 4513-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...