1-(4-Methoxyphenyl)-2-quinoxalin-2-ylethanone 95+%
Dodavatel: Apollo Scientific
OR7943-10G
OR7943-5G
OR7943-1G
APOSOR7943-10GEA
17260
CZK
APOSOR7943-10G
APOSOR7943-5G
APOSOR7943-1G
1-(4-Methoxyphenyl)-2-quinoxalin-2-ylethanone 95+%
1-(4-Methoxyphenyl)-2-quinoxalin-2-ylethanone
Vzorec:
C₁₇H₁₄N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD04037913 CAS číslo: 73100-63-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...