1-(4-tert-Butylphenyl)-2-pyrimidin-4-ylethanone 95+%
Dodavatel: Apollo Scientific
OR7939-25G
OR7939-5G
OR7939-1G
APOSOR7939-25GEA
35710
CZK
APOSOR7939-25G
APOSOR7939-5G
APOSOR7939-1G
1-(4-tert-Butylphenyl)-2-pyrimidin-4-ylethanone 95+%
1-(4-tert-Butylphenyl)-2-pyrimidin-4-ylethanone
Vzorec:
C₁₆H₁₈N₂O MW: 254,33 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04037907 CAS číslo: 849021-29-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...