Tisk…

3-tert-Butoxypropanenitrile

Dodavatel: Apollo Scientific
Varování

OR7930-1G OR7930-5G
APOSOR7930-1GEA 3610 CZK
APOSOR7930-1G APOSOR7930-5G
3-tert-Butoxypropanenitrile
3-tert-Butoxypropanenitrile
Vzorec: C₇H₁₃NO
MW: 127,19 g/mol
Bod varu: 62 °C/6,75mmHg
Storage Temperature: Ambient
MDL Number: MFCD04037897
CAS číslo: 99764-73-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR