Tisk…

4-(4-Chlorophenoxy)phenylhydrazine hydrochloride 95+%

Dodavatel: Apollo Scientific
Varování

OR7918-250MG OR7918-500MG
APOSOR7918-250MGEA 744 CZK
APOSOR7918-250MG APOSOR7918-500MG
4-(4-Chlorophenoxy)phenylhydrazine hydrochloride 95+%
4-(4-Chlorophenoxy)phenylhydrazine hydrochloride
Vzorec: C₁₂H₁₂Cl₂N₂O
Bod tání: 206…208 °C
Storage Temperature: Ambient
MDL Number: MFCD03426045
CAS číslo: 849021-09-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR