Tisk…

N-Benzylaminoacetaldehyde diethyl acetal

Dodavatel: Apollo Scientific
Varování

OR7899-25G OR7899-5G
APOSOR7899-25GEA 11540 CZK
APOSOR7899-25G APOSOR7899-5G
N-Benzylaminoacetaldehyde diethyl acetal
N-Benzylaminoacetaldehyde diethyl acetal
Vzorec: C₁₃H₂₁NO₂
MW: 223.32 g/mol
Bod varu: 90…92 °C/0.05mmHg
Storage Temperature: Ambient
MDL Number: MFCD00051599
CAS číslo: 61190-10-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR