Tisk…

4-(4,6-Dimethoxypyrimidin-2-yl)aniline 95+%

Dodavatel: Apollo Scientific
Nebezpečí

OR7881-1G OR7881-5G
APOSOR7881-1GEA 5960 CZK
APOSOR7881-1G APOSOR7881-5G
4-(4,6-Dimethoxypyrimidin-2-yl)aniline 95+%
4-(4,6-Dimethoxypyrimidin-2-yl)aniline
Vzorec: C₁₂H₁₃N₃O₂
Bod tání: 103…105 °C
Storage Temperature: Ambient
MDL Number: MFCD01319482
CAS číslo: 387350-86-9
UN: 2811
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR