Tisk…

3-Methylsulphonylphenylacetic acid

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-Methylsulphonylphenylacetic Acid

OR7865-1G OR7865-250MG
APOSOR7865-1GEA 8370 CZK
APOSOR7865-1G APOSOR7865-250MG
3-Methylsulphonylphenylacetic acid
3-Methylsulphonylphenylacetic acid
Vzorec: C₉H₁₀O₄S
MW: 214.24 g/mol
Bod tání: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD03426066
CAS číslo: 1877-64-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR