Tisk…

4-(3,4-Dihydropyrido[4,3-b]-1,6-naphthyridin-2(1H)-yl)-4-oxobutanoic acid 95+%

Dodavatel: Apollo Scientific

OR7841-1G OR7841-5G
APOSOR7841-1GEA 8370 CZK
APOSOR7841-1G APOSOR7841-5G
4-(3,4-Dihydropyrido[4,3-b]-1,6-naphthyridin-2(1H)-yl)-4-oxobutanoic acid 95+%
4-(3,4-Dihydropyrido[4,3-b]-1,6-naphthyridin-2(1H)-yl)-4-oxobutanoic acid
Vzorec: C₁₅H₁₅N₃O₃
MW: 285,31 g/mol
Bod tání: 221…224 °C
Storage Temperature: Ambient
MDL Number: MFCD00203194
CAS číslo: 392233-82-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR