3-(tert-Butoxycarbonyl)benzoic acid
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
mono-(tert-Butyl) isophthalate
OR7828-1G
OR7828-5G
OR7828-10G
APOSOR7828-1GEA
2400
CZK
APOSOR7828-1G
APOSOR7828-5G
APOSOR7828-10G
3-(tert-Butoxycarbonyl)benzoic acid
3-(tert-Butoxycarbonyl)benzoic acid
Vzorec:
C₁₂H₁₄O₄ MW: 222,24 g/mol Bod tání: 152…155 °C Storage Temperature: Ambient |
MDL Number:
MFCD04037899 CAS číslo: 33704-19-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...