1-(5-Bromopyrimidin-2-yl)piperidine-4-carboxylic acid 95+%
Dodavatel: Apollo Scientific
OR7826-1G
APOSOR7826-1GEA
4800
CZK
APOSOR7826-1G
1-(5-Bromopyrimidin-2-yl)piperidine-4-carboxylic acid 95+%
1-(5-Bromopyrimidin-2-yl)piperidine-4-carboxylic acid
Vzorec:
C₁₀H₁₂BrN₃O₂ Bod tání: 164…166 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426070 CAS číslo: 799283-92-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...