Tisk…

8-Bromo-1,6-naphthyridine-2-carboxylic acid 95+%

Dodavatel: Apollo Scientific
Varování

OR7824-1G OR7824-250MG
APOSOR7824-1GEA 15510 CZK
APOSOR7824-1G APOSOR7824-250MG
8-Bromo-1,6-naphthyridine-2-carboxylic acid 95+%
8-Bromo-1,6-naphthyridine-2-carboxylic acid
Vzorec: C₉H₅BrN₂O₂
Storage Temperature: Ambient
MDL Number: MFCD04037919
CAS číslo: 197507-55-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR