Tisk…

5-Bromo-2-methoxy-4-(methylthio)pyrimidine

Dodavatel: Apollo Scientific

OR7802-1G OR7802-5G
APOSOR7802-1GEA 10350 CZK
APOSOR7802-1G APOSOR7802-5G
5-Bromo-2-methoxy-4-(methylthio)pyrimidine
5-Bromo-2-methoxy-4-(methylthio)pyrimidine
Vzorec: C₆H₇BrN₂OS
Storage Temperature: Ambient
MDL Number: MFCD07781219
CAS číslo: 59549-52-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR