1-[4-(Methylsulphonyl)phenyl]piperazine 95+%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
1-(4-(Methylsulphonyl)phenyl)piperazine
OR7799-1G
OR7799-25G
OR7799-5G
APOSOR7799-1GEA
1780
CZK
APOSOR7799-1G
APOSOR7799-25G
APOSOR7799-5G
1-[4-(Methylsulphonyl)phenyl]piperazine 95+%
1-[4-(Methylsulphonyl)phenyl]piperazine
Vzorec:
C₁₁H₁₆N₂O₂S MW: 240,33 g/mol Bod tání: 148…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD03648601 CAS číslo: 187669-60-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...