4-Aminoindole
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
Indol-4-ylamine, 4-Indolamine, 4-Amino-1H-indole
OR7782-1G
OR7782-5G
APOSOR7782-1GEA
2280
CZK
APOSOR7782-1G
APOSOR7782-5G
4-Aminoindole
4-Aminoindole
Vzorec:
C₈H₈N₂ MW: 132,16 g/mol Bod tání: 106…108 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD01076559 CAS číslo: 5192-23-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...