Tisk…

N-[3-Aminomethyl)phenyl]acetamide hydrochloride 95+%

Dodavatel: Apollo Scientific
Varování

Synonyms: 3-(Aminomethyl)acetanilide hydrochloride

OR7774-1G OR7774-250MG
APOSOR7774-1GEA 3580 CZK
APOSOR7774-1G APOSOR7774-250MG
N-[3-Aminomethyl)phenyl]acetamide hydrochloride 95+%
N-[3-Aminomethyl)phenyl]acetamide hydrochloride
Vzorec: C₉H₁₃ClN₂O
MW: 200,67 g/mol
Bod tání: 230…233 °C
Storage Temperature: Ambient
MDL Number: MFCD04039171
CAS číslo: 238428-27-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR