4-Nitro-1,8-naphthalic anhydride 95%
Dodavatel: Apollo Scientific
|
Varování
|
Synonyms:
6-Nitrobenzo[de]isochromene-1,3-dione
OR7744-25G
OR7744-5G
APOSOR7744-25GEA
19680
CZK
APOSOR7744-25G
APOSOR7744-5G
4-Nitro-1,8-naphthalic anhydride 95%
4-Nitro-1,8-naphthalic anhydride
Vzorec:
C₁₂H₅NO₅ MW: 243,1718 g/mol Bod tání: 226…229 °C Storage Temperature: Ambient |
MDL Number:
MFCD00013446 CAS číslo: 6642-29-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...