Tisk…

4-(Dimethylamino)benzylamine 97%

Dodavatel: Apollo Scientific
Nebezpečí

Synonyms: 4-(Aminomethyl)-N,N-dimethylaniline

OR7726-1G OR7726-5G
APOSOR7726-1GEA 622 CZK
APOSOR7726-1G APOSOR7726-5G
4-(Dimethylamino)benzylamine 97%
4-(Dimethylamino)benzylamine
Vzorec: C₉H₁₄N₂
MW: 150,23 g/mol
Bod varu: 82…84 °C/0,8mmHg
Storage Temperature: Ambient
MDL Number: MFCD00466364
CAS číslo: 19293-58-4
UN: 2735
ADR: 8,II

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR